Assay Details
Target Gene Details
Entrez Gene ID: | 10047 |
Gene Name: | cystatin 8 |
Gene Aliases: |
CRES, CTES5 |
Location: |
Chr.20:23491077-23507124 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CST8 | NM_001281730.1 | NP_001268659.1 | ||
| NM_005492.3 | NP_005483.1 | |||
| AF059244.1 | AAC14707.1 | |||
| AW665128.1 | ||||
| BC069496.1 | AAH69496.1 | |||
| BC105113.1 | AAI05114.1 | |||
| BC105119.1 | AAI05120.1 | |||
| BX096109.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv458949 | Chr.20:23446011 - 23588763 on Build GRCh38 | Gain |
|
| esv3645525 | Chr.20:23425607 - 23597079 on Build GRCh38 | Gain |
|
| nsv1065390 | Chr.20:23432253 - 23588601 on Build GRCh38 | Loss |
|
| nsv1063655 | Chr.20:23416012 - 23509093 on Build GRCh38 | Gain |
|
| nsv585722 | Chr.20:23444001 - 23588763 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map