Assay Details
Target Gene Details
Entrez Gene ID: | 55245 |
Gene Name: | ubiquinol-cytochrome c reductase complex assembly factor 1 |
Gene Aliases: |
BFZB, C20orf44, CBP3, UQCC |
Location: |
Chr.20:35302566-35412142 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UQCC1 | NM_001184977.1 | NP_001171906.1 | ||
| NM_018244.4 | NP_060714.3 | |||
| NM_199487.2 | NP_955781.2 | |||
| XM_011528877.1 | XP_011527179.1 | |||
| XM_011528878.1 | XP_011527180.1 | |||
| XM_011528879.1 | XP_011527181.1 | |||
| XM_011528880.1 | XP_011527182.1 | |||
| XM_011528881.2 | XP_011527183.1 | |||
| XM_011528882.1 | XP_011527184.1 | |||
| XM_011528883.1 | XP_011527185.1 | |||
| XM_011528884.1 | XP_011527186.1 | |||
| AK001712.1 | BAA91854.1 | |||
| AK022650.1 | BAB14156.1 | |||
| AK022742.1 | BAB14217.1 | |||
| AK093804.1 | 1 | 262 | ||
| AK128252.1 | 1 | 1342 | ||
| AK225281.1 | ||||
| AK225557.1 | ||||
| AK293149.1 | ||||
| AK293516.1 | ||||
| AK295379.1 | ||||
| AK302133.1 | ||||
| AY050659.1 | AAL13118.1 | |||
| AY050660.1 | AAL13119.1 | |||
| BC105791.1 | AAI05792.1 | |||
| BC119013.1 | ||||
| BC122862.1 | ||||
| BM919736.1 | ||||
| BX537568.1 | CAD97785.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059168 | Chr.20:35217473 - 35319358 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map