Assay Details
Target Gene Details
Entrez Gene ID: | 3170 |
Gene Name: | forkhead box A2 |
Gene Aliases: |
HNF3B, TCF3B |
Location: |
Chr.20:22581004-22585463 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXA2 | NM_153675.2 | NP_710141.1 | ||
| AB028021.1 | BAA78106.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1073050 | Chr.20:22583661 - 22584862 on Build GRCh38 | Deletion |
|
| nsv1136325 | Chr.20:22583662 - 22584862 on Build GRCh38 | Deletion |
|
| nsv953003 | Chr.20:22575463 - 22586762 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map