Assay Details
Target Gene Details
Entrez Gene ID: | 10621 |
Gene Name: | RNA polymerase III subunit F |
Gene Aliases: |
RPC39, RPC6 |
Location: |
Chr.20:18467389-18484648 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR3F | NM_001282526.1 | NP_001269455.1 | ||
| NM_006466.3 | NP_006457.2 | |||
| NR_104209.1 | ||||
| XM_011529141.1 | XP_011527443.1 | |||
| XM_017027596.1 | XP_016883085.1 | |||
| AK096684.1 | ||||
| AK223298.1 | BAD97018.1 | |||
| AK290892.1 | ||||
| AK314838.1 | ||||
| BC012588.1 | AAH12588.1 | |||
| BC016761.1 | ||||
| DA729767.1 | ||||
| U93869.1 | AAB63677.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645437 | Chr.20:18357770 - 18484726 on Build GRCh38 | Gain |
|
| dgv4283n100 | Chr.20:18364565 - 18647377 on Build GRCh38 | Loss |
|
| nsv1062244 | Chr.20:18184227 - 18520274 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map