Assay Details
Target Gene Details
Entrez Gene ID: | 84515 |
Gene Name: | minichromosome maintenance 8 homologous recombination repair factor |
Gene Aliases: |
C20orf154, POF10, dJ967N21.5 |
Location: |
Chr.20:5950652-6000941 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCM8 | XM_017028105.1 | 19 | 8166 | XP_016883594.1 |
| XM_017028107.1 | 12 | 8256 | XP_016883596.1 |
Target Gene Details
Entrez Gene ID: | 101929225 |
Gene Name: | MCM8 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.20:5990795-6005821 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCM8-AS1 | NR_110101.1 | |||
| AK058185.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575356 | Chr.20:5954200 - 6068502 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map