Assay Details
Target Gene Details
Entrez Gene ID: | 85366 |
Gene Name: | myosin light chain kinase 2 |
Gene Aliases: |
KMLC, MLCK, MLCK2, skMLCK |
Location: |
Chr.20:31819375-31834697 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYLK2 | NM_033118.3 | NP_149109.1 | ||
| AF325549.1 | AAK15494.1 | |||
| AJ272502.1 | CAC81354.1 | |||
| BC069627.1 | AAH69627.1 | |||
| BC092413.1 | AAH92413.1 | |||
| BC127622.1 | ||||
| BX647635.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4300n100 | Chr.20:31247234 - 32033468 on Build GRCh38 | Gain |
|
| esv3645657 | Chr.20:31820268 - 31822444 on Build GRCh38 | Loss |
|
| esv2664840 | Chr.20:31820468 - 31822431 on Build GRCh38 | Deletion |
|
| esv2676878 | Chr.20:31821092 - 31823589 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map