Assay Details
Target Gene Details
Entrez Gene ID: | 631 |
Gene Name: | beaded filament structural protein 1 |
Gene Aliases: |
CP115, CP94, CTRCT33, LIFL-H |
Location: |
Chr.20:17493905-17569220 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BFSP1 | NM_001161705.1 | NP_001155177.1 | ||
| NM_001195.4 | NP_001186.1 | |||
| NM_001278606.1 | NP_001265535.1 | |||
| NM_001278607.1 | NP_001265536.1 | |||
| NM_001278608.1 | NP_001265537.1 | |||
| XM_017028005.1 | XP_016883494.1 | |||
| AF039655.1 | AAB94939.1 | |||
| AK304673.1 | ||||
| AK316247.1 | ||||
| BC041483.1 | AAH41483.1 | |||
| BQ230198.1 | ||||
| Y16717.2 | CAA76348.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760670 | Chr.20:17316866 - 17555369 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map