Assay Details
Target Gene Details
Entrez Gene ID: | 10406 |
Gene Name: | WAP four-disulfide core domain 2 |
Gene Aliases: |
EDDM4, HE4, WAP5, dJ461P17.6 |
Location: |
Chr.20:45469754-45481532 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WFDC2 | NM_006103.3 | NP_006094.3 | ||
| AF330259.1 | AAL37485.1 | |||
| AF330260.1 | AAL37486.1 | |||
| AF330262.1 | AAL37488.1 | |||
| AK225528.1 | ||||
| AK290288.1 | 3 | 509 | ||
| AK311966.1 | ||||
| AY212888.1 | AAO52683.1 | |||
| BC046106.1 | AAH46106.1 | |||
| BI759775.1 | ||||
| CR456977.1 | CAG33258.1 | |||
| X63187.1 | CAA44869.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map