Assay Details
Target Gene Details
Entrez Gene ID: | 51604 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class T |
Gene Aliases: |
CGI-06, MCAHS3, NDAP, PNH2 |
Location: |
Chr.20:45416067-45426245 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
| dgv4319n100 | Chr.20:45384898 - 45428463 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map