Assay Details
Target Gene Details
Entrez Gene ID: | 101927159 |
Gene Name: | uncharacterized LOC101927159 |
Gene Aliases: |
- |
Location: |
Chr.20:42685483-42688562 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927159 | NR_110004.1 |
Target Gene Details
Entrez Gene ID: | 11122 |
Gene Name: | protein tyrosine phosphatase, receptor type T |
Gene Aliases: |
RPTPrho |
Location: |
Chr.20:42072752-43189917 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPRT | NM_007050.5 | NP_008981.4 | ||
| NM_133170.3 | NP_573400.3 | |||
| XM_017027611.1 | XP_016883100.1 | |||
| XM_017027612.1 | XP_016883101.1 | |||
| XM_017027613.1 | XP_016883102.1 | |||
| AB006621.2 | ||||
| AF043644.4 | AAD09421.2 | |||
| BC153300.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057847 | Chr.20:42652464 - 44431352 on Build GRCh38 | Loss |
|
| esv3893336 | Chr.20:42329007 - 43025374 on Build GRCh38 | Loss |
|
| dgv782e214 | Chr.20:42655671 - 42715083 on Build GRCh38 | Loss |
|
| nsv833988 | Chr.20:42599148 - 42751281 on Build GRCh38 | Loss |
|
| nsv586028 | Chr.20:42668354 - 42752575 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map