Assay Details
Target Gene Details
Entrez Gene ID: | 23394 |
Gene Name: | activity dependent neuroprotector homeobox |
Gene Aliases: |
ADNP1, HVDAS, MRD28 |
Location: |
Chr.20:50888918-50934984 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADNP | XM_017027757.1 | 1 | 2328 | XP_016883246.1 |
Target Gene Details
Entrez Gene ID: | 101927631 |
Gene Name: | ADNP antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.20:50930984-50945134 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADNP-AS1 | NR_110007.1 | |||
| NR_110008.1 | ||||
| NR_110009.1 | ||||
| AI827739.1 | ||||
| BF064247.1 | ||||
| HY015168.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834002 | Chr.20:50810235 - 50986764 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map