Assay Details
Target Gene Details
Entrez Gene ID: | 58495 |
Gene Name: | ovo like zinc finger 2 |
Gene Aliases: |
EUROIMAGE566589, PPCD1, ZNF339 |
Location: |
Chr.20:18024152-18059188 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OVOL2 | NM_001303461.1 | NP_001290390.1 | ||
| NM_001303462.1 | NP_001290391.1 | |||
| NM_021220.3 | NP_067043.2 | |||
| AK022284.1 | BAB14002.1 | |||
| AL079276.1 | CAB45151.1 | |||
| BC006148.1 | AAH06148.1 | |||
| BE791524.1 | ||||
| BM563561.1 | ||||
| BT007295.1 | AAP35959.1 | |||
| BX331154.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833933 | Chr.20:18029399 - 18206602 on Build GRCh38 | Gain |
|
| nsv458945 | Chr.20:18005414 - 18038630 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map