Assay Details
Target Gene Details
Entrez Gene ID: | 5770 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 1 |
Gene Aliases: |
PTP1B |
Location: |
Chr.20:50510321-50584762 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPN1 | NM_001278618.1 | NP_001265547.1 | ||
| NM_002827.3 | NP_002818.1 | |||
| AK290638.1 | ||||
| AK299830.1 | ||||
| AK316563.1 | ||||
| AU117677.1 | ||||
| BC015660.2 | AAH15660.1 | |||
| BC018164.1 | AAH18164.1 | |||
| BT006752.1 | AAP35398.1 | |||
| DC300354.1 | ||||
| M31724.1 | AAA60223.1 | |||
| M33689.1 | AAA60157.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4324n100 | Chr.20:50512568 - 50535619 on Build GRCh38 | Gain |
|
| dgv163n111 | Chr.20:50509110 - 50534089 on Build GRCh38 | Duplication |
|
| nsv586177 | Chr.20:50358658 - 50546698 on Build GRCh38 | Gain |
|
| esv2760682 | Chr.20:50493794 - 50535631 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map