Assay Details
Target Gene Details
Entrez Gene ID: | 57419 |
Gene Name: | solute carrier family 24 member 3 |
Gene Aliases: |
NCKX3 |
Location: |
Chr.20:19212646-19722897 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC24A3 | NM_020689.3 | NP_065740.2 | ||
| AF169257.2 | AAG12988.2 | |||
| AF288087.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73122902,rs73124804] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map