Assay Details
Target Gene Details
Entrez Gene ID: | 9139 |
Gene Name: | CBFA2/RUNX1 translocation partner 2 |
Gene Aliases: |
EHT, MTGR1, ZMYND3, p85 |
Location: |
Chr.20:33490068-33650031 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CBFA2T2 | NM_001032999.2 | NP_001028171.1 | ||
| XM_017028122.1 | XP_016883611.1 | |||
| AK294168.1 | ||||
| AK307887.1 | ||||
| CB998035.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064413 | Chr.20:33348381 - 33599978 on Build GRCh38 | Gain |
|
| dgv4302n100 | Chr.20:33413503 - 33558912 on Build GRCh38 | Loss |
|
| nsv522690 | Chr.20:33427230 - 33552481 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map