Assay Details
Target Gene Details
Entrez Gene ID: | 3240 |
Gene Name: | haptoglobin |
Gene Aliases: |
BP, HP2ALPHA2, HPA1S |
Location: |
Chr.16:72054592-72061056 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HP | NM_001126102.2 | NP_001119574.1 | ||
| NM_001318138.1 | NP_001305067.1 | |||
| NM_005143.4 | NP_005134.1 | |||
| AK055872.1 | 1 | 1087 | ||
| AK314700.1 | ||||
| BC017862.1 | ||||
| BC058031.1 | AAH58031.1 | |||
| BC070299.1 | AAH70299.1 | |||
| BC107587.1 | AAI07588.1 | |||
| BC121124.1 | ||||
| BC121125.1 | ||||
| BJ994335.1 | ||||
| CB117006.1 | ||||
| CB134891.1 | ||||
| K00422.1 | AAA52687.1 | |||
| K01763.1 | AAA52684.1 | |||
| L29394.1 | AAA52685.1 | |||
| X00637.1 | CAA25267.1 |
Target Gene Details
Entrez Gene ID: | 54957 |
Gene Name: | thioredoxin like 4B |
Gene Aliases: |
DLP, Dim2 |
Location: |
Chr.16:72044261-72094316 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TXNL4B | XM_017023377.1 | XP_016878866.1 | ||
| AK308891.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv504e199 | Chr.16:72049451 - 72076250 on Build GRCh38 | Deletion |
|
| esv2714683 | Chr.16:71950823 - 72151945 on Build GRCh38 | Deletion |
|
| nsv833273 | Chr.16:71937902 - 72118955 on Build GRCh38 | Loss |
|
| dgv3017n100 | Chr.16:72040933 - 72757730 on Build GRCh38 | Gain |
|
| dgv530e214 | Chr.16:72046969 - 72065087 on Build GRCh38 | Gain |
|
| esv2478287 | Chr.16:72054766 - 72064976 on Build GRCh38 | Deletion |
|
| esv2215660 | Chr.16:72054699 - 72063557 on Build GRCh38 | Deletion |
|
| nsv978167 | Chr.16:72054423 - 72084772 on Build GRCh38 | Duplication |
|
| nsv827729 | Chr.16:72049820 - 72076352 on Build GRCh38 | Loss |
|
| nsv428329 | Chr.16:71942441 - 72240448 on Build GRCh38 | Gain |
|
| nsv9456 | Chr.16:72050862 - 72084423 on Build GRCh38 | Gain+Loss |
|
| esv2422047 | Chr.16:72053111 - 72074194 on Build GRCh38 | Duplication |
|
| esv23639 | Chr.16:72054601 - 72078398 on Build GRCh38 | Gain+Loss |
|
| nsv817737 | Chr.16:72054432 - 72074194 on Build GRCh38 | Loss |
|
| esv2714685 | Chr.16:72054836 - 72063593 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map