Assay Details
Target Gene Details
Entrez Gene ID: | 404734 |
Gene Name: | ANKHD1-EIF4EBP3 readthrough |
Gene Aliases: |
MASK-BP3, MASK-BP3ARF |
Location: |
Chr.5:140401814-140549578 on Build GRCh38 |
Assay Gene Location: | Within Intron 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKHD1-EIF4EBP3 | NM_020690.5 | NP_065741.3 | ||
| AB029008.1 | BAA83037.1 | |||
| AF521883.1 | AAO14944.1 | |||
| AK125734.1 |
Target Gene Details
Entrez Gene ID: | 8637 |
Gene Name: | eukaryotic translation initiation factor 4E binding protein 3 |
Gene Aliases: |
4E-BP3, 4EBP3 |
Location: |
Chr.5:140547666-140549578 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF4EBP3 | NM_003732.2 | NP_003723.1 | ||
| AF038869.1 | AAC39761.1 | |||
| BC010881.1 | AAH10881.1 | |||
| BC069293.1 | AAH69293.1 | |||
| BC073751.1 | AAH73751.1 |
More Information
Set Membership: |
|

Genomic Map