Assay Details
Target Gene Details
Entrez Gene ID: | 338758 |
Gene Name: | long intergenic non-protein coding RNA 936 |
Gene Aliases: |
- |
Location: |
Chr.12:89708955-89711952 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC00936 | NR_028138.1 | 1 | 857 | |
AK094517.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv559705 | Chr.12:89702019 - 89713058 on Build GRCh38 | Gain |
![]() ![]() |
nsv559709 | Chr.12:89708471 - 89713058 on Build GRCh38 | Gain |
![]() ![]() |
