Assay Details
Target Gene Details
Entrez Gene ID: | 768239 |
Gene Name: | prosaposin-like 1 (gene/pseudogene) |
Gene Aliases: |
- |
Location: |
Chr.4:7430294-7434973 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSAPL1 | NM_001085382.1 | 1 | 2860 | NP_001078851.1 |
| DQ991252.1 | 1 | 2860 |
Target Gene Details
Entrez Gene ID: | 57537 |
Gene Name: | sortilin related VPS10 domain containing receptor 2 |
Gene Aliases: |
- |
Location: |
Chr.4:7192607-7742837 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SORCS2 | NM_020777.2 | NP_065828.2 | ||
| XM_005247987.4 | XP_005248044.2 | |||
| XM_011513514.2 | XP_011511816.1 | |||
| XM_011513515.2 | XP_011511817.1 | |||
| XM_011513516.2 | XP_011511818.1 | |||
| XM_017008481.1 | XP_016863970.1 | |||
| AF286190.1 | AAL04014.1 | |||
| AK295629.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv949781 | Chr.4:7412874 - 7473273 on Build GRCh38 | Deletion |
|
| nsv461203 | Chr.4:7422024 - 7432880 on Build GRCh38 | Loss |
|
| nsv593569 | Chr.4:7289636 - 7624409 on Build GRCh38 | Gain |
|
| nsv508994 | Chr.4:7206788 - 7502929 on Build GRCh38 | Insertion |
|
| nsv1003679 | Chr.4:7317384 - 7447535 on Build GRCh38 | Gain |
|
| nsv461204 | Chr.4:7426172 - 7436625 on Build GRCh38 | Loss |
|
| nsv469758 | Chr.4:7368356 - 7544523 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map