Assay Details
Target Gene Details
Entrez Gene ID: | 256987 |
Gene Name: | serine incorporator 5 |
Gene Aliases: |
C5orf12, TPO1 |
Location: |
Chr.5:80111227-80256082 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERINC5 | NM_001174071.2 | NP_001167542.1 | ||
| NM_178276.6 | NP_840060.1 | |||
| NR_126060.1 | ||||
| NR_126061.1 | ||||
| AF498273.1 | AAP06800.1 | |||
| BC101280.2 | AAI01281.1 | |||
| BC101283.2 | AAI01284.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3605549 | Chr.5:80068073 - 80166801 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79245378] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map