Assay Details
Target Gene Details
Entrez Gene ID: | 105377934 |
Gene Name: | putative uncharacterized protein C6orf183 |
Gene Aliases: |
- |
Location: |
Chr.6:109165792-109203801 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105377934 | XM_017011615.1 | XP_016867104.1 | ||
| XM_017011616.1 | XP_016867105.1 | |||
| XM_017011617.1 | XP_016867106.1 | |||
| XM_017011618.1 | XP_016867107.1 | |||
| XM_017011619.1 | XP_016867108.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs139124725] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map