Assay Details
Target Gene Details
Entrez Gene ID: | 285753 |
Gene Name: | centrosomal protein 57 like 1 |
Gene Aliases: |
C6orf182, bA487F23.2, cep57R |
Location: |
Chr.6:109095130-109167696 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEP57L1 | XM_005266935.4 | 11 | 4623 | XP_005266992.3 |
| XM_005266936.4 | 11 | 4623 | XP_005266993.3 | |
| XM_005266937.4 | 11 | 4575 | XP_005266994.3 | |
| XM_005266938.4 | 11 | 4572 | XP_005266995.3 | |
| XM_006715462.3 | 12 | 4987 | XP_006715525.1 | |
| XM_011535759.2 | 11 | 4937 | XP_011534061.1 | |
| XM_011535762.2 | 9 | 4211 | XP_011534064.1 | |
| XM_017010764.1 | 12 | 5126 | XP_016866253.1 | |
| XM_017010765.1 | 13 | 5381 | XP_016866254.1 | |
| XM_017010767.1 | 11 | 4937 | XP_016866256.1 | |
| XM_017010768.1 | 11 | 4751 | XP_016866257.1 | |
| XM_017010769.1 | 11 | 4639 | XP_016866258.1 | |
| XM_017010770.1 | 13 | 5330 | XP_016866259.1 | |
| XM_017010771.1 | 9 | 4216 | XP_016866260.1 | |
| XM_017010772.1 | 10 | 4675 | XP_016866261.1 |
Target Gene Details
Entrez Gene ID: | 105377934 |
Gene Name: | putative uncharacterized protein C6orf183 |
Gene Aliases: |
- |
Location: |
Chr.6:109165792-109203801 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105377934 | XM_017011615.1 | XP_016867104.1 | ||
| XM_017011616.1 | XP_016867105.1 | |||
| XM_017011617.1 | XP_016867106.1 | |||
| XM_017011618.1 | XP_016867107.1 | |||
| XM_017011619.1 | XP_016867108.1 |
More Information
Set Membership: |
|

Genomic Map