Assay Details
Target Gene Details
Entrez Gene ID: | 105377967 |
Gene Name: | uncharacterized LOC105377967 |
Gene Aliases: |
- |
Location: |
Chr.6:118085903-118095472 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105377967 | NR_134601.1 | |||
| DB526410.1 |
Target Gene Details
Entrez Gene ID: | 222553 |
Gene Name: | solute carrier family 35 member F1 |
Gene Aliases: |
C6orf169, dJ230I3.1 |
Location: |
Chr.6:117907226-118317676 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC35F1 | NM_001029858.3 | NP_001025029.2 | ||
| XM_005266865.4 | XP_005266922.1 | |||
| AK295860.1 | ||||
| BC028615.1 | AAH28615.1 | |||
| BC036493.1 | AAH36493.1 | |||
| BC114925.1 | AAI14926.1 |
More Information
Set Membership: |
|

Genomic Map