Assay Details
Target Gene Details
Entrez Gene ID: | 441161 |
Gene Name: | oocyte expressed protein |
Gene Aliases: |
C6orf156, FLOPED, HOEP19, KHDC2 |
Location: |
Chr.6:73368553-73369888 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OOEP | NM_001080507.2 | NP_001073976.1 | ||
| XM_017010890.1 | XP_016866379.1 | |||
| BX093821.1 | ||||
| EU290647.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830693 | Chr.6:73339619 - 73506357 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map