Assay Details
Target Gene Details
Entrez Gene ID: | 729020 |
Gene Name: | ribulose-5-phosphate-3-epimerase like 1 |
Gene Aliases: |
- |
Location: |
Chr.10:103245887-103248016 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPEL1 | NM_001143909.1 | 1 | 209 | NP_001137381.1 |
| BC137242.1 | 1 | 148 | ||
| BC137243.1 | 1 | 148 | ||
| BC157829.1 | 1 | 148 | ||
| BC157875.1 | 1 | 209 | ||
| BC171806.1 | 1 | 148 | ||
| BC171885.1 | 1 | 209 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509365 | Chr.10:103207187 - 103316390 on Build GRCh38 | Insertion |
|
| nsv948175 | Chr.10:103245020 - 103247057 on Build GRCh38 | Duplication |
|
| esv3624406 | Chr.10:103167084 - 103248522 on Build GRCh38 | Gain |
|
| esv3624408 | Chr.10:103202044 - 103247021 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map