Assay Details
Target Gene Details
Entrez Gene ID: | 105 |
Gene Name: | adenosine deaminase, RNA specific B2 (inactive) |
Gene Aliases: |
ADAR3, RED2 |
Location: |
Chr.10:1177313-1737476 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADARB2 | NM_018702.3 | NP_061172.1 | ||
| AF034837.1 | AAF78094.1 | |||
| AI695657.1 | ||||
| BC137477.1 | ||||
| BC139725.1 | ||||
| BC140852.1 |
Target Gene Details
Entrez Gene ID: | 642394 |
Gene Name: | ADARB2 antisense RNA 1 |
Gene Aliases: |
C10orf109, NCRNA00168, bA466B20.1 |
Location: |
Chr.10:1526630-1556984 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADARB2-AS1 | NR_033387.1 | |||
| AF086274.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046041 | Chr.10:1149274 - 2146961 on Build GRCh38 | Gain |
|
| dgv925n54 | Chr.10:1198080 - 1862404 on Build GRCh38 | Gain |
|
| nsv1041498 | Chr.10:1071260 - 1717996 on Build GRCh38 | Gain |
|
| nsv1054555 | Chr.10:1515950 - 1922951 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114269924,rs76186670] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map