Assay Details
Target Gene Details
Entrez Gene ID: | 335 |
Gene Name: | apolipoprotein A1 |
Gene Aliases: |
apo(a) |
Location: |
Chr.11:116835751-116837950 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOA1 | NM_000039.2 | NP_000030.1 | ||
| NM_001318017.1 | NP_001304946.1 | |||
| NM_001318018.1 | NP_001304947.1 | |||
| NM_001318021.1 | NP_001304950.1 | |||
| A14829.1 | CAA01198.1 | |||
| A15879.1 | CAA01253.1 | |||
| AK292231.1 | ||||
| AL531353.3 | ||||
| AL557908.3 | ||||
| BC005380.1 | AAH05380.1 | |||
| BC110286.1 | AAI10287.1 | |||
| BU600916.1 | ||||
| CD558050.1 | ||||
| M11791.1 | AAA35545.1 | |||
| M27875.1 | AAA62829.1 | |||
| M29068.1 | AAA51747.1 | |||
| T68127.1 | ||||
| X00566.1 | CAA25232.1 | |||
| X02162.1 | CAA26097.1 |
Target Gene Details
Entrez Gene ID: | 104326055 |
Gene Name: | APOA1 antisense RNA |
Gene Aliases: |
- |
Location: |
Chr.11:116836117-116855729 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOA1-AS | NR_126362.1 | |||
| DA327409.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs7116797] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map