Assay Details
Target Gene Details
Entrez Gene ID: | 101929089 |
Gene Name: | uncharacterized LOC101929089 |
Gene Aliases: |
- |
Location: |
Chr.11:118511918-118531094 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929089 | NR_120576.1 | |||
| AW611756.1 |
Target Gene Details
Entrez Gene ID: | 143941 |
Gene Name: | tetratricopeptide repeat domain 36 |
Gene Aliases: |
HBP21 |
Location: |
Chr.11:118527495-118531025 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC36 | NM_001080441.1 | 3 | 507 | NP_001073910.1 |
| BC146916.1 | 3 | 530 | ||
| BC146921.1 | 3 | 530 | ||
| BC171906.1 | 4 | 642 | ||
| BC171913.1 | 4 | 665 | ||
| EU489483.1 | 3 | 569 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044281 | Chr.11:118503814 - 118554704 on Build GRCh38 | Gain |
|
| esv3627872 | Chr.11:118502100 - 118536457 on Build GRCh38 | Gain |
|

Genomic Map