Assay Details
Target Gene Details
Entrez Gene ID: | 123624 |
Gene Name: | ATP/GTP binding protein like 1 |
Gene Aliases: |
CCP4, FECD8 |
Location: |
Chr.15:86079635-87031476 on Build GRCh38 |
Assay Gene Location: | Within Intron 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGBL1 | XM_011521227.2 | XP_011519529.1 | ||
| DA695799.1 |
Target Gene Details
Entrez Gene ID: | 102724452 |
Gene Name: | uncharacterized LOC102724452 |
Gene Aliases: |
- |
Location: |
Chr.15:86938797-86988426 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724452 | NR_135683.1 | |||
| BX100197.1 |
Target Gene Details
Entrez Gene ID: | 105370954 |
Gene Name: | uncharacterized LOC105370954 |
Gene Aliases: |
- |
Location: |
Chr.15:86932880-86946331 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105370954 | NR_135682.1 | |||
| AA833610.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039254 | Chr.15:86527126 - 87399757 on Build GRCh38 | Loss |
|
| nsv519797 | Chr.15:86084875 - 87238541 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map