Assay Details
Target Gene Details
Entrez Gene ID: | 100128731 |
Gene Name: | oligosaccharyltransferase complex subunit 4, non-catalytic |
Gene Aliases: |
- |
Location: |
Chr.2:27070472-27071699 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OST4 | NM_001134693.1 | NP_001128165.1 | ||
| AA772256.1 | ||||
| AL049447.1 | ||||
| BF697310.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6744n54 | Chr.2:26973431 - 27151141 on Build GRCh38 | Loss |
|
| nsv817673 | Chr.2:27037601 - 27163931 on Build GRCh38 | Gain |
|
| nsv833603 | Chr.2:27059106 - 27263631 on Build GRCh38 | Loss |
|
| nsv455974 | Chr.2:26989775 - 27128411 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map