Assay Details
Target Gene Details
Entrez Gene ID: | 10865 |
Gene Name: | AT-rich interaction domain 5A |
Gene Aliases: |
MRF-1, MRF1, RP11-363D14 |
Location: |
Chr.2:96536719-96552638 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARID5A | NM_001319085.1 | NP_001306014.1 | ||
| NM_001319087.1 | NP_001306016.1 | |||
| NM_001319093.1 | NP_001306022.1 | |||
| NM_001319094.1 | NP_001306023.1 | |||
| NM_001319096.1 | NP_001306025.1 | |||
| NM_212481.2 | NP_997646.1 | |||
| XM_017003184.1 | XP_016858673.1 | |||
| AK300173.1 | ||||
| AK304566.1 | ||||
| AK307768.1 | ||||
| AY358167.1 | AAQ88534.1 | |||
| BM556343.1 | ||||
| BX396215.2 | ||||
| BX648918.1 | ||||
| CR981009.1 | ||||
| HY002073.1 | ||||
| HY076710.1 | ||||
| M62324.1 | AAA36325.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2720413 | Chr.2:96472591 - 96793459 on Build GRCh38 | Deletion |
|
| dgv607e201 | Chr.2:95855884 - 97561622 on Build GRCh38 | Deletion |
|
| nsv1135873 | Chr.2:95981454 - 97579645 on Build GRCh38 | Deletion |
|
| esv3425717 | Chr.2:95859900 - 97515905 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114244789] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map