Assay Details
Target Gene Details
Entrez Gene ID: | 643314 |
Gene Name: | KIAA0754 |
Gene Aliases: |
- |
Location: |
Chr.1:39409504-39416482 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA0754 | NM_015038.1 | 1 | 1272 | NP_055853.1 |
Target Gene Details
Entrez Gene ID: | 23499 |
Gene Name: | microtubule-actin crosslinking factor 1 |
Gene Aliases: |
ABP620, ACF7, MACF, OFC4 |
Location: |
Chr.1:39084167-39487138 on Build GRCh38 |
Assay Gene Location: | Within Intron 66 |
More Information
Additional Information:
For this assay, SNP(s) [rs77885848] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map