Assay Details
Target Gene Details
Entrez Gene ID: | 56997 |
Gene Name: | coenzyme Q8A |
Gene Aliases: |
ADCK3, ARCA2, CABC1, COQ10D4, COQ8, SCAR9 |
Location: |
Chr.1:226939339-226987545 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COQ8A | NM_020247.4 | NP_064632.2 | ||
| XM_005273201.1 | XP_005273258.1 | |||
| XM_011544238.1 | XP_011542540.1 | |||
| XM_011544239.2 | XP_011542541.1 | |||
| XM_011544240.2 | XP_011542542.1 | |||
| XM_011544241.2 | XP_011542543.1 | |||
| XM_017001852.1 | XP_016857341.1 | |||
| AB073905.1 | BAB91363.1 | |||
| AK074693.1 | BAC11143.1 | |||
| AK090494.1 | ||||
| AK092784.1 | ||||
| AK126200.1 | ||||
| AK293571.1 | ||||
| AK297735.1 | ||||
| AK300537.1 | ||||
| AK301012.1 | ||||
| BC005171.2 | AAH05171.2 | |||
| BC129930.1 | ||||
| BX648860.1 | CAH56132.1 |
Target Gene Details
Entrez Gene ID: | 105379282 |
Gene Name: | glucose-dependent insulinotropic receptor-like |
Gene Aliases: |
- |
Location: |
Chr.1:226971536-226977439 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105379282 | XM_011544346.2 | 2 | 3834 | XP_011542648.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952131 | Chr.1:226973700 - 226988299 on Build GRCh38 | Deletion |
|
| nsv520923 | Chr.1:226939917 - 226991247 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map