Assay Details
Target Gene Details
Entrez Gene ID: | 84301 |
Gene Name: | DNA damage inducible 1 homolog 2 |
Gene Aliases: |
- |
Location: |
Chr.1:15617458-15661057 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDI2 | NM_032341.4 | 10 | 2123 | NP_115717.3 |
| AK057374.1 | 8 | 1519 |
Target Gene Details
Entrez Gene ID: | 6248 |
Gene Name: | regulatory solute carrier protein, family 1, member 1 |
Gene Aliases: |
RS1 |
Location: |
Chr.1:15659713-15662030 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RSC1A1 | NM_006511.2 | 1 | 788 | NP_006502.1 |
| AK314753.1 | 1 | 788 | ||
| CR999543.1 | 1 | 563 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006877 | Chr.1:15642115 - 15707706 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map