Assay Details
Target Gene Details
Entrez Gene ID: | 10715 |
Gene Name: | ceramide synthase 1 |
Gene Aliases: |
EPM8, LAG1, LASS1, UOG1 |
Location: |
Chr.19:18868546-18896727 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CERS1 | NM_021267.4 | NP_067090.1 |
Target Gene Details
Entrez Gene ID: | 2657 |
Gene Name: | growth differentiation factor 1 |
Gene Aliases: |
DORV, DTGA3, RAI |
Location: |
Chr.19:18868546-18896144 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GDF1 | NM_001492.5 | NP_001483.3 | ||
| M62302.1 | AAA58501.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv828473 | Chr.19:18864321 - 18884066 on Build GRCh38 | Gain |
|
| nsv470132 | Chr.19:18853633 - 18943911 on Build GRCh38 | Loss |
|
| nsv1151217 | Chr.19:18627369 - 19284914 on Build GRCh38 | Deletion |
|
| nsv833777 | Chr.19:18765126 - 18920267 on Build GRCh38 | Loss |
|
| nsv952423 | Chr.19:18859292 - 18941091 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map