Assay Details
Target Gene Details
Entrez Gene ID: | 6703 |
Gene Name: | small proline rich protein 2D |
Gene Aliases: |
- |
Location: |
Chr.1:153039725-153041144 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPRR2D | NM_006945.4 | NP_008876.3 | ||
| XM_017002175.1 | XP_016857664.1 | |||
| AK291317.1 | ||||
| BC120938.1 | ||||
| BG183546.1 | ||||
| M21302.1 | AAA36640.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv547899 | Chr.1:152699529 - 153272606 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map