Assay Details
Target Gene Details
Entrez Gene ID: | 399687 |
Gene Name: | myosin XVIIIA |
Gene Aliases: |
MYSPDZ, SPR210 |
Location: |
Chr.17:29073510-29180389 on Build GRCh38 |
Assay Gene Location: | Within Exon 42 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYO18A | NM_078471.3 | 42 | 7111 | NP_510880.2 |
| NM_203318.1 | 41 | 7066 | NP_976063.1 | |
| AB177858.1 | 41 | 6930 | BAD66836.1 | |
| AB177860.1 | 41 | 6018 | BAD66838.1 | |
| AF252258.1 | 4 | 1364 | AAF70181.1 | |
| AK122870.1 | 8 | 2202 | ||
| AK303522.1 | ||||
| AY703984.1 | 41 | 6018 | AAV80770.1 | |
| BC039612.1 | 40 | 7059 | AAH39612.1 | |
| CR933614.1 | 41 | 7041 | CAI45931.1 | |
| D86970.2 | 42 | 7134 |
Target Gene Details
Entrez Gene ID: | 9220 |
Gene Name: | TGFB1-induced anti-apoptotic factor 1 |
Gene Aliases: |
MAJN, SPR210 |
Location: |
Chr.17:29073521-29075609 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TIAF1 | NM_004740.3 | 1 | 1628 | NP_004731.2 |
| AF105277.1 | 1 | 337 | AAD04172.1 | |
| AK054958.1 | 1 | 1628 | ||
| AK126861.1 | 1 | 3326 | ||
| BC047669.1 | 1 | 925 | AAH47669.1 | |
| BC058075.1 | 1 | 611 | AAH58075.1 | |
| BC131561.1 | 1 | 825 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833408 | Chr.17:28913470 - 29105450 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map