Assay Details
Target Gene Details
Entrez Gene ID: | 79648 |
Gene Name: | microcephalin 1 |
Gene Aliases: |
BRIT1, MCT |
Location: |
Chr.8:6406592-6648505 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCPH1 | NM_001322042.1 | NP_001308971.1 | ||
| NM_024596.4 | NP_078872.2 | |||
| XM_011534755.2 | XP_011533057.1 | |||
| XM_011534760.2 | XP_011533062.1 | |||
| XM_017013829.1 | XP_016869318.1 | |||
| XM_017013830.1 | XP_016869319.1 | |||
| XM_017013831.1 | XP_016869320.1 | |||
| XM_017013832.1 | XP_016869321.1 | |||
| XM_017013833.1 | XP_016869322.1 | |||
| XM_017013834.1 | XP_016869323.1 | |||
| AI480294.1 | ||||
| AK022909.1 | BAB14304.1 |
Target Gene Details
Entrez Gene ID: | 100507530 |
Gene Name: | MCPH1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.8:6615603-6708209 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCPH1-AS1 | NR_125386.1 | 5 | 3458 | |
| AK057771.1 | 1 | 922 | BAB71565.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1021684 | Chr.8:6492570 - 6951933 on Build GRCh38 | Loss |
|
| nsv1161706 | Chr.8:6602062 - 6630317 on Build GRCh38 | Duplication |
|
| nsv610004 | Chr.8:6544845 - 6915391 on Build GRCh38 | Gain |
|
| nsv1025976 | Chr.8:6615427 - 6829450 on Build GRCh38 | Loss |
|
| nsv6058 | Chr.8:6599506 - 6616816 on Build GRCh38 | Insertion |
|
| esv2422383 | Chr.8:6504868 - 6924071 on Build GRCh38 | Duplication |
|
| esv3616094 | Chr.8:6548175 - 6638852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map