Assay Details
Target Gene Details
Entrez Gene ID: | 23331 |
Gene Name: | tetratricopeptide repeat domain 28 |
Gene Aliases: |
TPRBK |
Location: |
Chr.22:27978014-28679865 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC28 | NM_001145418.1 | NP_001138890.1 | ||
| XM_005261405.2 | XP_005261462.1 | |||
| XM_006724171.3 | XP_006724234.1 | |||
| XM_011530018.2 | XP_011528320.1 | |||
| XM_017028673.1 | XP_016884162.1 | |||
| AB028966.3 | BAA82995.3 | |||
| AB665284.1 | ||||
| BC016465.1 | AAH16465.1 |
Target Gene Details
Entrez Gene ID: | 284900 |
Gene Name: | TTC28 antisense RNA 1 |
Gene Aliases: |
TTC28-AS, TTC28AS, dJ353E16.2 |
Location: |
Chr.22:27919376-28002679 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC28-AS1 | NR_026963.1 | 4 | 1385 | |
| AB051435.1 | BAB33318.1 | |||
| AL832597.1 | 4 | 1340 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3568347 | Chr.22:27995410 - 28002579 on Build GRCh38 | Loss |
|
| nsv522306 | Chr.22:27982484 - 28006636 on Build GRCh38 | Loss |
|
| esv3568348 | Chr.22:27986182 - 28002208 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map