Assay Details
Target Gene Details
Entrez Gene ID: | 25769 |
Gene Name: | solute carrier family 24 member 2 |
Gene Aliases: |
NCKX2 |
Location: |
Chr.9:19507452-20307888 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC24A2 | NM_001193288.2 | 9 | 4615 | NP_001180217.1 |
| NM_020344.3 | 10 | 4666 | NP_065077.1 | |
| XM_005251425.2 | 11 | 4945 | XP_005251482.1 | |
| XM_005251426.4 | 11 | 5477 | XP_005251483.1 | |
| XM_006716750.3 | 10 | 5426 | XP_006716813.1 | |
| XM_017014592.1 | 15 | 5416 | XP_016870081.1 | |
| XM_017014593.1 | 10 | 4898 | XP_016870082.1 | |
| AK127092.1 | 1 | 118 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv613734 | Chr.9:19470633 - 19533874 on Build GRCh38 | Gain |
|
| nsv613732 | Chr.9:19419530 - 19594813 on Build GRCh38 | Gain |
|
| nsv831524 | Chr.9:19440356 - 19622086 on Build GRCh38 | Gain+Loss |
|
| nsv466286 | Chr.9:19281101 - 19516865 on Build GRCh38 | Gain |
|
| esv3619913 | Chr.9:19433835 - 19551230 on Build GRCh38 | Gain |
|
| nsv524474 | Chr.9:19430333 - 19583439 on Build GRCh38 | Gain |
|
| esv3619911 | Chr.9:19379247 - 19517379 on Build GRCh38 | Gain |
|
| nsv1015645 | Chr.9:19469848 - 19570511 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map