Assay Details
Target Gene Details
Entrez Gene ID: | 285877 |
Gene Name: | POM121 transmembrane nucleoporin like 12 |
Gene Aliases: |
- |
Location: |
Chr.7:53035656-53036925 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POM121L12 | NM_182595.3 | 1 | 822 | NP_872401.3 |
| AL833888.1 | 1 | 473 | CAD38744.1 | |
| BX104052.1 | 1 | 127 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1021212 | Chr.7:51605303 - 53572805 on Build GRCh38 | Gain |
|
| nsv5745 | Chr.7:53034041 - 53067277 on Build GRCh38 | Insertion |
|
| nsv831001 | Chr.7:52935841 - 53085090 on Build GRCh38 | Loss |
|
| nsv1017676 | Chr.7:52864446 - 53282831 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map