Assay Details
Target Gene Details
Entrez Gene ID: | 55971 |
Gene Name: | BAI1 associated protein 2 like 1 |
Gene Aliases: |
IRTKS |
Location: |
Chr.7:98291650-98401115 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BAIAP2L1 | NM_018842.4 | NP_061330.2 | ||
| AF119666.2 | AAF17223.2 | |||
| AK027109.1 | ||||
| AK027142.1 | BAB15671.1 | |||
| AK074419.1 | ||||
| AK124269.1 | 1 | 646 | ||
| BC013888.2 | AAH13888.1 |
Target Gene Details
Entrez Gene ID: | 25798 |
Gene Name: | brain protein I3 |
Gene Aliases: |
I3 |
Location: |
Chr.7:98281667-98322398 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRI3 | XM_017011931.1 | XP_016867420.1 | ||
| XM_017011932.1 | XP_016867421.1 | |||
| XM_017011933.1 | XP_016867422.1 | |||
| XM_017011934.1 | XP_016867423.1 | |||
| XM_017011935.1 | XP_016867424.1 | |||
| XM_017011936.1 | XP_016867425.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831067 | Chr.7:98230857 - 98436216 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map