Assay Details
Target Gene Details
Entrez Gene ID: | 100101267 |
Gene Name: | POM121 transmembrane nucleoporin C |
Gene Aliases: |
POM121-2 |
Location: |
Chr.7:75416781-75486288 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POM121C | NM_001099415.2 | NP_001092885.2 | ||
| AK293781.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain |
|
| nsv981535 | Chr.7:75393440 - 75515890 on Build GRCh38 | Duplication |
|
| nsv831035 | Chr.7:75322437 - 75436287 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map