Assay Details
Target Gene Details
Entrez Gene ID: | 64983 |
Gene Name: | mitochondrial ribosomal protein L32 |
Gene Aliases: |
HSPC283, L32mt, MRP-L32, bMRP-59b |
Location: |
Chr.7:42932205-42937857 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL32 | NM_031903.2 | NP_114109.1 | ||
| AB049649.1 | BAB40854.1 | |||
| AF161401.1 | AAF28961.1 | |||
| AK090551.1 | 2 | 478 | ||
| AK313393.1 | ||||
| BC013147.1 | AAH13147.1 | |||
| BM833110.1 | ||||
| BU687817.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv606715 | Chr.7:42169735 - 42940343 on Build GRCh38 | Gain |
|
| nsv830979 | Chr.7:42806983 - 42987012 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map