Assay Details
Target Gene Details
Entrez Gene ID: | 5478 |
Gene Name: | peptidylprolyl isomerase A |
Gene Aliases: |
CYPA, CYPH, HEL-S-69p |
Location: |
Chr.7:44796636-44803123 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPIA | NM_001300981.1 | NP_001287910.1 | ||
| NM_021130.4 | NP_066953.1 | |||
| AB451307.1 | ||||
| AB451438.1 | ||||
| AK130101.1 | ||||
| AK290085.1 | ||||
| AK290851.1 | ||||
| AK293003.1 | ||||
| AK297340.1 | ||||
| AK316564.1 | ||||
| AU100120.1 | ||||
| BC093076.1 | AAH93076.1 | |||
| BC137057.1 | ||||
| BC137058.1 | ||||
| BM806224.1 | ||||
| BU786475.1 | ||||
| CD014106.1 | ||||
| CR456707.1 | CAG32988.1 | |||
| Y00052.1 | CAA68264.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830985 | Chr.7:44793475 - 44979588 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map