Assay Details
Target Gene Details
Entrez Gene ID: | 10320 |
Gene Name: | IKAROS family zinc finger 1 |
Gene Aliases: |
CVID13, Hs.54452, IK1, IKAROS, LYF1, LyF-1, PPP1R92, PRO0758, ZNFN1A1 |
Location: |
Chr.7:50303453-50405101 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IKZF1 | NM_001291839.1 | NP_001278768.1 | ||
| XM_011515058.2 | XP_011513360.1 | |||
| XM_011515059.2 | XP_011513361.1 | |||
| XM_011515061.2 | XP_011513363.1 | |||
| XM_011515063.2 | 1 | 214 | XP_011513365.1 | |
| XM_011515065.2 | XP_011513367.1 | |||
| XM_011515067.2 | XP_011513369.1 | |||
| XM_011515068.2 | 1 | 214 | XP_011513370.1 | |
| XM_011515070.2 | XP_011513372.1 | |||
| XM_011515071.2 | XP_011513373.1 | |||
| XM_011515072.2 | XP_011513374.1 | |||
| XM_011515074.2 | XP_011513376.1 | |||
| XM_011515076.2 | 1 | 241 | XP_011513378.1 | |
| XM_011515077.2 | XP_011513379.1 | |||
| XM_011515078.2 | XP_011513380.1 | |||
| XM_017011667.1 | 1 | 214 | XP_016867156.1 | |
| XM_017011668.1 | XP_016867157.1 | |||
| XM_017011669.1 | XP_016867158.1 | |||
| XM_017011670.1 | 1 | 172 | XP_016867159.1 | |
| DC391353.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950213 | Chr.7:50304305 - 50331004 on Build GRCh38 | Duplication |
|
| nsv1076892 | Chr.7:50303703 - 50304504 on Build GRCh38 | Deletion |
|
| esv27276 | Chr.7:50303611 - 50304811 on Build GRCh38 | Loss |
|
| nsv606897 | Chr.7:49632287 - 50387038 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map