Assay Details
Target Gene Details
Entrez Gene ID: | 389362 |
Gene Name: | proteasome assembly chaperone 4 |
Gene Aliases: |
C6orf86, PAC4, bA506K6.2 |
Location: |
Chr.6:3254325-3308224 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMG4 | XM_011514591.2 | 1 | 344 | XP_011512893.1 |
| XM_011514595.2 | XP_011512897.1 | |||
| AK125314.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv10275n54 | Chr.6:3254222 - 3255032 on Build GRCh38 | Gain |
|
| esv29057 | Chr.6:3253683 - 3262320 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map