Assay Details
Target Gene Details
Entrez Gene ID: | 3087 |
Gene Name: | hematopoietically expressed homeobox |
Gene Aliases: |
HEX, HMPH, HOX11L-PEN, PRH, PRHX |
Location: |
Chr.10:92689924-92695651 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HHEX | NM_002729.4 | NP_002720.1 | ||
| AK309463.1 | ||||
| AK314891.1 | ||||
| BC014336.1 | AAH14336.1 | |||
| BC015110.1 | AAH15110.1 | |||
| BC050638.1 | AAH50638.1 | |||
| DB504532.1 | ||||
| L16499.1 | AAA02988.1 | |||
| X67235.1 | CAA47661.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv168n67 | Chr.10:92637392 - 92712752 on Build GRCh38 | Gain |
|
| dgv1350n54 | Chr.10:92689403 - 92690681 on Build GRCh38 | Gain |
|
| nsv551939 | Chr.10:92689930 - 92690681 on Build GRCh38 | Gain+Loss |
|
| nsv551938 | Chr.10:92689860 - 92690681 on Build GRCh38 | Loss |
|
| dgv1352n54 | Chr.10:92689583 - 92690681 on Build GRCh38 | Gain+Loss |
|
| nsv1046258 | Chr.10:92107212 - 92706349 on Build GRCh38 | Gain |
|
| esv29580 | Chr.10:92689451 - 92690728 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map