Assay Details
Target Gene Details
Entrez Gene ID: | 10160 |
Gene Name: | FERM, ARH/RhoGEF and pleckstrin domain protein 1 |
Gene Aliases: |
CDEP, FARP1-IT1, PLEKHC2, PPP1R75 |
Location: |
Chr.13:98142562-98449773 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FARP1 | NM_001001715.3 | NP_001001715.2 | ||
| NM_001286839.1 | NP_001273768.1 | |||
| NM_005766.3 | NP_005757.1 | |||
| XM_011521046.2 | XP_011519348.1 | |||
| XM_017020312.1 | XP_016875801.1 | |||
| AB008430.1 | BAA24267.1 | |||
| AI273567.1 | ||||
| AI274149.1 | ||||
| BC041595.1 | AAH41595.1 | |||
| BC071592.1 | AAH71592.1 | |||
| BM763139.1 | ||||
| CF994038.1 | ||||
| DB498534.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3343n54 | Chr.13:98142835 - 98144218 on Build GRCh38 | Loss |
|
| nsv1118269 | Chr.13:98142946 - 98144046 on Build GRCh38 | Deletion |
|
| dgv3344n54 | Chr.13:98143027 - 98143998 on Build GRCh38 | Gain |
|
| nsv562860 | Chr.13:98142784 - 98144218 on Build GRCh38 | Gain |
|
| dgv3342n54 | Chr.13:98142480 - 98143998 on Build GRCh38 | Gain |
|
| esv27463 | Chr.13:98142463 - 98144115 on Build GRCh38 | Loss |
|
| esv2751157 | Chr.13:98089059 - 98147329 on Build GRCh38 | Loss |
|
| nsv562866 | Chr.13:98143221 - 98144052 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map