Assay Details
Target Gene Details
Entrez Gene ID: | 150946 |
Gene Name: | GRB2 associated regulator of MAPK1 subtype 2 |
Gene Aliases: |
FAM59B, GAREML |
Location: |
Chr.2:26173091-26202405 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAREM2 | NM_001168241.1 | NP_001161713.1 | ||
| XM_006711951.3 | XP_006712014.1 | |||
| XM_011532567.2 | XP_011530869.1 | |||
| AB124552.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv22083 | Chr.2:26172742 - 26173461 on Build GRCh38 | Loss |
|
| nsv1071583 | Chr.2:26172830 - 26173331 on Build GRCh38 | Deletion |
|
| nsv524918 | Chr.2:26169872 - 26193167 on Build GRCh38 | Gain |
|
| esv33316 | Chr.2:25440017 - 26672800 on Build GRCh38 | Loss |
|

Genomic Map